Benzyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Supplier: Matrix Scientific
003086-1G
003086-5G
101785-194EA
446.15
USD
101785-194
101785-192
Benzyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Benzyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
CAS: 175137-29-8
MDL Number: MFCD00068148
Molecular Formula: C13H8ClF3N2O2
Molecular Weight: 316,67
Hazard: Irritant
Formula:
C₁₃H₈ClF₃N₂O₂ MW: 316.67 g/mol Melting Pt: 45…47 °C |
MDL Number:
MFCD00068148 CAS Number: 175137-29-8 |
Specification Test Results
Melting point | 45-47°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...