3'-Hydroxybenzanilide

Supplier: Matrix Scientific

003005-5G
101785-052EA 81.08 USD
101785-052
3'-Hydroxybenzanilide
3'-Hydroxybenzanilide

CAS: 3743-28-0
MDL Number: MFCD00068823
Molecular Formula: C13H11NO2
Molecular Weight: 213,24
Hazard: Irritant

Order Now

Specification Test Results

Melting point 153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR