5-Ethoxycarbonyl-4-(trifluoromethyl)-pyrimidin-2-(1H)-one, Technical Grade
Supplier: Matrix Scientific
Synonyms:
Ethyl 1,2-dihydro-2-oxo-6-(trifluoromethyl)pyrimidine-5-carboxylate
002988-5G
002988-1G
101785-010EA
103.08
USD
101785-010
101785-008
5-Ethoxycarbonyl-4-(trifluoromethyl)-pyrimidin-2-(1H)-one, Technical Grade
5-Ethoxycarbonyl-4-(trifluoromethyl)-pyrimidin-2-(1H)-one
CAS: 154934-97-1
MDL Number: MFCD00052077
Molecular Formula: C8H7F3N2O3
Molecular Weight: 236,15
Hazard: Irritant
Formula:
C₈H₇F₃N₂O₃ MW: 236.15 g/mol Melting Pt: 176…178 °C |
MDL Number:
MFCD00052077 CAS Number: 154934-97-1 |
Specification Test Results
Melting point | 176-178°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...