Methyl-3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate
Supplier: Matrix Scientific
002971-1G
002971-5G
101784-984EA
90.77
USD
101784-984
101784-986
Methyl-3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate
Methyl-3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate
CAS: 175137-08-3
MDL Number: MFCD00068133
Molecular Formula: C12H10FNO2S
Molecular Weight: 251,28
Hazard: Irritant
Formula:
C₁₂H₁₀FNO₂S MW: 251.28 g/mol Melting Pt: 153…155 °C |
MDL Number:
MFCD00068133 CAS Number: 175137-08-3 |
Specification Test Results
Melting point | 153-155°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...