DL-Mandelamide ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Hydroxy-2-phenylacetamide
002968-5G
002968-25G
101784-976EA
130.77
USD
101784-976
101784-978
DL-Mandelamide ≥98%
DL-Mandelamide
Purity: 98%
CAS: 4410-31-5
MDL Number: MFCD00025495
Molecular Formula: C8H9NO2
Molecular Weight: 151,17
Hazard: Irritant
Formula:
C₈H₉NO₂ MW: 151.17 g/mol Melting Pt: 133…134 °C |
MDL Number:
MFCD00025495 CAS Number: 4410-31-5 |
Specification Test Results
Melting point | 133-134°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...