Methyl-2-aminothiophene-3-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 2-aminothiophene-3-carboxylate

029254-25G
101827-784EA 110.77 USD
101827-784
Methyl-2-aminothiophene-3-carboxylate
Methyl 2-aminothiophene-3-carboxylate

Hazard: Irritant

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Melting Pt: 76…81 °C
MDL Number: MFCD00159547
CAS Number: 4651-81-4

Order Now

Specification Test Results

Melting point 76-81 ºC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR