Methyl-2-amino-5-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-5-methylthiophene-3-carboxylate

028908-500MG
101827-090EA 15.38 USD
101827-090
Methyl-2-amino-5-methylthiophene-3-carboxylate
Methyl-2-amino-5-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Melting Pt: 113…118 °C
MDL Number: MFCD01922101
CAS Number: 19369-53-0

Order Now

Specification Test Results

Melting point 113-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR