3-Aminopicolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-2-pyridinecarboxylic acid3-Aminopyridine-2-carboxylic acid2-Pyridinecarboxylic acid, 3-amino-

018356-5G
101806-032EA 104.62 USD
101806-032
3-Aminopicolinic acid ≥95%
3-Aminopicolinic acid

Hazard: Irritant

Formula: C₆H₆N₂O₂
MW: 138.126 g/mol
Melting Pt: 218…222 °C
MDL Number: MFCD00090153
CAS Number: 1462-86-8

Order Now

Specification Test Results

Melting point 218-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR