2-Iodo-6-methoxypyridine ≥98%

Supplier: Matrix Scientific

018309-250MG
101805-890EA 320 USD
101805-890
2-Iodo-6-methoxypyridine ≥98%
2-Iodo-6-methoxypyridine

Hazard: Irritant

Formula: C₆H₆INO
MW: 235.0200 g/mol
Melting Pt: 39…40 °C
MDL Number: MFCD07366733
CAS Number: 182275-70-3

Order Now

Specification Test Results

Melting point 39-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR