2-Bromo-3-iodoisonicotinic acid ≥98%

Supplier: Matrix Scientific

018306-500MG
101805-882EA 196.92 USD
101805-882
2-Bromo-3-iodoisonicotinic acid ≥98%
2-Bromo-3-iodoisonicotinic acid

Hazard: Irritant

Formula: C₆H₃BrINO₂
Melting Pt: 214…215 °C
MDL Number: MFCD07366731
CAS Number: 848243-29-8

Order Now

Specification Test Results

Melting point 214-215°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR