5-Bromo-4,6-dichloropyrimidine ≥97%
Supplier: Matrix Scientific
018271-1G
101805-774EA
35.38
USD
101805-774
5-Bromo-4,6-dichloropyrimidine ≥97%
5-Bromo-4,6-dichloropyrimidine
Hazard: Toxic/Corrosive/Stored under Argon/Light Sensitive
Formula:
C₄HBrCl₂N₂ Melting Pt: 77…81 °C |
MDL Number:
MFCD00047379 CAS Number: 68797-61-5 |
Specification Test Results
Melting point | 77-81°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...