4-Amino-3-chlorobenzonitrile ≥98%

Supplier: Matrix Scientific

018264-25G
101805-766EA 75.38 USD
101805-766
4-Amino-3-chlorobenzonitrile ≥98%
4-Amino-3-chlorobenzonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 100…102 °C
MDL Number: MFCD00052917
CAS Number: 21803-75-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR