3-Iodo-2-(2,2,2-trimethylacetamido)pyridine

Supplier: Matrix Scientific

Synonyms: N-(3-Iodopyridin-2-yl)pivalamideN-(3-Iod-2-pyridyl)pivalamide

018226-1G
101805-680EA 159.85 USD
101805-680
3-Iodo-2-(2,2,2-trimethylacetamido)pyridine
N-(3-Iod-2-pyridyl)pivalamide

Hazard: Irritant

Formula: C₁₀H₁₃IN₂O
MW: 304.13 g/mol
Melting Pt: 148.8…149.0 °C
MDL Number: MFCD05662384
CAS Number: 113975-31-8

Order Now

Specification Test Results

Melting point 148.8-149.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR