1-[4-Fluoro-2-(methylsulfonyl)phenyl]piperazine

Supplier: Matrix Scientific

009346-1G
101796-912EA 230.77 USD
101796-912
1-[4-Fluoro-2-(methylsulfonyl)phenyl]piperazine
1-[4-Fluoro-2-(methylsulfonyl)phenyl]piperazine

Hazard: Irritant

Formula: C₁₁H₁₅FN₂O₂S
Melting Pt: 153…155 °C
MDL Number: MFCD04037948
CAS Number: 849938-78-9

Order Now

Specification Test Results

Melting point 153-155°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR