5-Fluoro-2-(methylsulfonyl)benzoic acid

Supplier: Matrix Scientific

009335-1G
101796-890EA 120.75 USD
101796-890
5-Fluoro-2-(methylsulfonyl)benzoic acid
5-Fluoro-2-(methylsulphonyl)benzoic acid

Hazard: Irritant

Formula: C₈H₇FO₄S
Melting Pt: 147…149 °C
MDL Number: MFCD04037937
CAS Number: 773873-55-5

Order Now

Specification Test Results

Melting point 147-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR