1,2-Dichloro-4-methylsulfonylbenzene

Supplier: Matrix Scientific

009331-1G
101796-880EA 29.23 USD
101796-880
1,2-Dichloro-4-methylsulfonylbenzene
1,2-Dichloro-4-methylsulphonylbenzene

Hazard: Irritant

Formula: C₇H₆Cl₂O₂S
Melting Pt: 109…110 °C
MDL Number: MFCD00025063
CAS Number: 38452-47-0

Order Now

Specification Test Results

Melting point 109-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR