2,6-Dichloroisonicotinamide

Supplier: Matrix Scientific

009274-5G 009274-1G
101796-758EA 235.38 USD
101796-758 101796-756
2,6-Dichloroisonicotinamide
2,6-Dichloroisonicotinamide

Hazard: Irritant

Formula: C₆H₄Cl₂N₂O
MW: 191.0200 g/mol
Melting Pt: 212…214 °C
MDL Number: MFCD00052640
CAS Number: 89281-13-0

Order Now

Specification Test Results

Melting point 202-206°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR