4-(Trifluoromethoxy)phenylhydrazine hydrochloride ≥98%

Supplier: Matrix Scientific

Synonyms: (4-(Trifluoromethoxy)phenyl)hydrazine hydrochloride

009198-5G 009198-25G
101796-624EA 67.85 USD
101796-624 101796-622
4-(Trifluoromethoxy)phenylhydrazine hydrochloride ≥98%
4-(Trifluoromethoxy)phenylhydrazine hydrochloride

Hazard: Irritant

Formula: C₇H₈ClF₃N₂O
MW: 228.60 g/mol
Melting Pt: 240…243 °C
MDL Number: MFCD00053033
CAS Number: 133115-72-7

Order Now

Specification Test Results

Melting point 240-243°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR