4-Bromo-2-fluorobenzylamine hydrochloride ≥98%
Supplier: Matrix Scientific
Synonyms:
(4-Bromo-2-fluorophenyl)methanamine hydrochloride
008838-25G
008838-5G
101795-572EA
24.62
USD
101795-572
101795-574
4-Bromo-2-fluorobenzylamine hydrochloride ≥98%
4-Bromo-2-fluorobenzylamine hydrochloride
Hazard: Irritant
Formula:
C₇H₇BrFN·HCl MW: 240.50 g/mol Melting Pt: 254…257 °C |
MDL Number:
MFCD00143266 CAS Number: 147181-08-6 |
Specification Test Results
Melting point | 254-257°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...