6-Chloro-1H-pyrrolo[3,2-c]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-5-azaindole

058638-1G
101915-198EA 347.69 USD
101915-198
6-Chloro-1H-pyrrolo[3,2-c]pyridine ≥95%
6-Chloro-1H-pyrrolo[3,2-c]pyridine

Hazard: Irritant

Formula: C₇H₅ClN₂
MW: 152.5800 g/mol
Melting Pt: 190…191 °C
MDL Number: MFCD08690133
CAS Number: 74976-31-1

Order Now

Specification Test Results

Melting point 190-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR