4-Bromo-2-(trifluoromethoxy)benzoic acid ≥98%

Supplier: Matrix Scientific

058518-1G 058518-5G
101915-022EA 23.08 USD
101915-022 101915-024
4-Bromo-2-(trifluoromethoxy)benzoic acid ≥98%
4-Bromo-2-(trifluoromethoxy)benzoic acid

Hazard: Irritant

Formula: C₈H₄BrF₃O₃
MW: 285.0200 g/mol
Melting Pt: 91…94 °C
MDL Number: MFCD12546979
CAS Number: 509142-48-7

Order Now

Specification Test Results

Melting point 91-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR