2-(2-Oxo-1,3-oxazolan-3-yl)ethyl4-methylbenzenesulfonate ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(2-Oxo-1,3-oxazolidin-3-yl)ethyl toluene-4-sulphonate
045526-1G
045526-500MG
045526-5G
101871-302EA
806.15
USD
101871-302
101871-298
101871-300
2-(2-Oxo-1,3-oxazolan-3-yl)ethyl4-methylbenzenesulfonate ≥95%
2-(2-Oxo-1,3-oxazolan-3-yl)ethyl4-methylbenzenesulfonate
Hazard: Irritant
Formula:
C₁₂H₁₅NO₅S MW: 285.32 g/mol Melting Pt: 68…69 °C |
MDL Number:
MFCD08276953 CAS Number: 159974-55-7 |
Specification Test Results
Melting point | 68-69°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...