2-(2-Thienyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one ≥95%

Supplier: Matrix Scientific

045523-1G 045523-5G 045523-500MG
101871-280EA 278.46 USD
101871-280 101871-282 101871-284
2-(2-Thienyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one ≥95%
2-(2-Thienyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one

Hazard: Irritant

Formula: C₁₃H₁₁NOS₂
Melting Pt: 159…161 °C
MDL Number: MFCD01927811
CAS Number: 5871-68-1

Order Now

Specification Test Results

Melting point 159-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR