5-Bromo-2-hydroxy-3-methylbenzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-hydroxy-3-methylbenzoic acid
045513-1G
045513-500MG
045513-5G
101871-236EA
532.31
USD
101871-236
101871-232
101871-234
5-Bromo-2-hydroxy-3-methylbenzenecarboxylic acid ≥95%
5-Bromo-2-hydroxy-3-methylbenzenecarboxylic acid
Hazard: Irritant
Formula:
C₈H₇BrO₃ MW: 231.05 g/mol Melting Pt: 231…234 °C |
MDL Number:
MFCD00093269 CAS Number: 36194-82-8 |
Specification Test Results
Melting point | 231-234°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...