Methyl-5-nitro-2-(1-pyrrolidinyl)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-nitro-2-pyrrolidin-1-ylbenzoate
045509-5G
045509-500MG
045509-1G
101871-218EA
806.15
USD
101871-218
101871-214
101871-216
Methyl-5-nitro-2-(1-pyrrolidinyl)benzenecarboxylate ≥95%
Methyl-5-nitro-2-(1-pyrrolidinyl)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₄ MW: 250.26 g/mol Melting Pt: 141…144 °C |
MDL Number:
MFCD00662491 CAS Number: 259269-68-6 |
Specification Test Results
Melting point | 141-144°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...