2-(4-Aminophenoxy)benzoic acid ≥97%
Supplier: Matrix Scientific
045502-5G
045502-1G
045502-500MG
101871-180EA
387.69
USD
101871-180
101871-182
101871-178
2-(4-Aminophenoxy)benzoic acid ≥97%
2-(4-Aminophenoxy)benzoic acid
Hazard: Irritant
Formula:
C₁₃H₁₁NO₃ Melting Pt: 180…182 °C |
MDL Number:
MFCD06659583 CAS Number: 67724-03-2 |
Specification Test Results
Melting point | 180-182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...