Methyl-4-(4-methoxyphenyl)-2,4-dioxobutanoate ≥95%
Supplier: Matrix Scientific
045495-5G
045495-1G
045495-500MG
101871-138EA
118.46
USD
101871-138
101871-140
101871-142
Methyl-4-(4-methoxyphenyl)-2,4-dioxobutanoate ≥95%
Methyl-4-(4-methoxyphenyl)-2,4-dioxobutanoate
Hazard: Irritant
Formula:
C₁₂H₁₂O₅ MW: 236.23 g/mol Melting Pt: 96…98 °C |
MDL Number:
MFCD04967380 CAS Number: 39757-31-8 |
Specification Test Results
Melting point | 96-98°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...