3-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(1,3,4,5,6,7-Hexahydro-3-oxo-2H-indazol-2-yl)benzoic acid
045488-5G
045488-500MG
045488-1G
101871-100EA
232.31
USD
101871-100
101871-102
101871-098
3-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid ≥95%
3-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₄N₂O₃ MW: 258.28 g/mol Melting Pt: 281 °C |
MDL Number:
MFCD06659578 CAS Number: 885949-86-0 |
Specification Test Results
Melting point | 281°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...