2-Butyl-5-nitrobenzoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(But-1-yl)-5-nitro-1,3-benzoxazole

045487-1G 045487-5G 045487-500MG
101871-092EA 278.46 USD
101871-092 101871-094 101871-096
2-Butyl-5-nitrobenzoxazole ≥95%
2-Butyl-5-nitrobenzoxazole

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
Melting Pt: 45…47 °C
MDL Number: MFCD05663810
CAS Number: 886360-98-1

Order Now

Specification Test Results

Melting point 45-47°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR