2-Butyl-6-nitrobenzoxazole ≥95%

Supplier: Matrix Scientific

045486-5G 045486-500MG 045486-1G
101871-090EA 806.15 USD
101871-090 101871-086 101871-088
2-Butyl-6-nitrobenzoxazole ≥95%
2-Butyl-6-nitrobenzoxazole

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
Melting Pt: 19…20 °C
MDL Number: MFCD05663809
CAS Number: 886360-96-9

Order Now

Specification Test Results

Melting point 19-20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR