6-Chloroimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

045481-500MG 045481-1G 045481-5G
101871-060EA 53.85 USD
101871-060 101871-056 101871-058
6-Chloroimidazo[1,2-a]pyridine ≥95%
6-Chloroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD05663807
CAS Number: 6188-25-6

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR