2-(2-Pyridinyl)-4-(trifluoromethyl)-5-pyrimidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

045430-1G 045430-500MG
101870-798EA 927.69 USD
101870-798 101870-796
2-(2-Pyridinyl)-4-(trifluoromethyl)-5-pyrimidinecarboxylic acid ≥95%
2-(2-Pyridinyl)-4-(trifluoromethyl)-5-pyrimidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₆F₃N₃O₂
MW: 269.18 g/mol
Melting Pt: 224…226 °C
MDL Number: MFCD06200983
CAS Number: 874816-10-1

Order Now

Specification Test Results

Melting point 224-226°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR