6-Methyl-2-(2,2,2-trifluoro-1-hydroxyethyl)-3-pyridinol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-2-(1-hydroxy-2,2,2-trifluoroethyl)-6-methylpyridine

045409-500MG 045409-5G 045409-1G
101870-690EA 806.15 USD
101870-690 101870-688 101870-686
6-Methyl-2-(2,2,2-trifluoro-1-hydroxyethyl)-3-pyridinol ≥95%
6-Methyl-2-(2,2,2-trifluoro-1-hydroxyethyl)-3-pyridinol

Hazard: Irritant

Formula: C₈H₈F₃NO₂
MW: 207.15 g/mol
Melting Pt: 138…141 °C
MDL Number: MFCD03425809
CAS Number: 344776-71-2

Order Now

Specification Test Results

Melting point 138-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR