2-[(3-Acetylanilino)carbonyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045400-500MG 045400-1G 045400-5G
101870-638EA 181.54 USD
101870-638 101870-642 101870-640
2-[(3-Acetylanilino)carbonyl]benzenecarboxylic acid ≥95%
2-[(3-Acetylanilino)carbonyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₃NO₄
Melting Pt: 189…191 °C
MDL Number: MFCD00090455
CAS Number: 194784-73-1

Order Now

Specification Test Results

Melting point 189-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR