1-(3-Acetylphenyl)-3,3-dimethyl-2-azetanone ≥95%
Supplier: Matrix Scientific
045392-5G
045392-500MG
045392-1G
101870-602EA
806.15
USD
101870-602
101870-600
101870-598
1-(3-Acetylphenyl)-3,3-dimethyl-2-azetanone ≥95%
1-(3-Acetylphenyl)-3,3-dimethyl-2-azetanone
Hazard: Irritant
Formula:
C₁₃H₁₅NO₂ MW: 217.27 g/mol Melting Pt: 92…94 °C |
MDL Number:
MFCD02570851 CAS Number: 453557-75-0 |
Specification Test Results
Melting point | 92-94°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...