3-[(tert-Butoxycarbonyl)amino]-3-(2-thienyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045371-500MG 045371-1G
101870-528EA 375.38 USD
101870-528 101870-530
3-[(tert-Butoxycarbonyl)amino]-3-(2-thienyl)propanoic acid ≥95%
3-[(tert-Butoxycarbonyl)amino]-3-(2-thienyl)propanoic acid

Hazard: Irritant

Formula: C₁₂H₁₇NO₄S
MW: 271.34 g/mol
Melting Pt: 99…102 °C
MDL Number: MFCD02090671
CAS Number: 53030-49-2

Order Now

Specification Test Results

Melting point 99-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR