Methyl-2-phenyl-3-(2-thienyl)acrylate ≥95%

Supplier: Matrix Scientific

045361-500MG 045361-1G 045361-5G
101870-500EA 806.15 USD
101870-500 101870-498 101870-496
Methyl-2-phenyl-3-(2-thienyl)acrylate ≥95%
Methyl-2-phenyl-3-(2-thienyl)acrylate

Hazard: Irritant

Formula: C₁₄H₁₂O₂S
Melting Pt: 81…83 °C
MDL Number: MFCD03001283
CAS Number: 30302-64-8

Order Now

Specification Test Results

Melting point 81-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR