3-Amino-3-(2-pyridinyl)acrylonitrile ≥95%
Supplier: Matrix Scientific
045352-5G
045352-1G
045352-500MG
101870-464EA
461.54
USD
101870-464
101870-462
101870-460
3-Amino-3-(2-pyridinyl)acrylonitrile ≥95%
3-Amino-3-(2-pyridinyl)acrylonitrile
Hazard: Irritant
Formula:
C₈H₇N₃ MW: 145.17 g/mol Melting Pt: 116…118 °C |
MDL Number:
MFCD01871647 CAS Number: 55330-52-4 |
Specification Test Results
Melting point | 116-118°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...