N-(4-Methylbenzyl)urea ≥95%

Supplier: Matrix Scientific

045332-5G 045332-500MG 045332-1G
101870-360EA 532.31 USD
101870-360 101870-358 101870-356
N-(4-Methylbenzyl)urea ≥95%
N-(4-Methylbenzyl)urea

Hazard: Irritant

Formula: C₉H₁₂N₂O
MW: 164.21 g/mol
Melting Pt: 198…199 °C
MDL Number: MFCD01314625
CAS Number: 54582-34-2

Order Now

Specification Test Results

Melting point 198-199°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR