6-Methyl-3-(2-thienyl)-1,2,4-triazin-5-ol ≥95%

Supplier: Matrix Scientific

045322-1G 045322-500MG 045322-5G
101870-300EA 1600 USD
101870-300 101870-298 101870-296
6-Methyl-3-(2-thienyl)-1,2,4-triazin-5-ol ≥95%
6-Methyl-3-(2-thienyl)-1,2,4-triazin-5-ol

Hazard: Irritant

Formula: C₈H₇N₃OS
Melting Pt: 292…294 °C
MDL Number: MFCD04117817
CAS Number: 886360-71-0

Order Now

Specification Test Results

Melting point 292-294°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR