6-Methylimidazo[2,1-b][1,3]thiazole-5-carbohydrazide ≥95%

Supplier: Matrix Scientific

045321-500MG
101870-294EA 278.46 USD
101870-294
6-Methylimidazo[2,1-b][1,3]thiazole-5-carbohydrazide ≥95%
6-Methylimidazo[2,1-b][1,3]thiazole-5-carbohydrazide

Hazard: Irritant

Formula: C₇H₈N₄OS
Melting Pt: 183…184 °C
MDL Number: MFCD03086213
CAS Number: 161563-79-7

Order Now

Specification Test Results

Melting point 183-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR