Methyl-4-(2-pyrimidinylamino)butanoate ≥95%
Supplier: Matrix Scientific
045318-5G
045318-1G
045318-500MG
101870-282EA
467.69
USD
101870-282
101870-280
101870-284
Methyl-4-(2-pyrimidinylamino)butanoate ≥95%
Methyl-4-(2-pyrimidinylamino)butanoate
Hazard: Irritant
Formula:
C₉H₁₃N₃O₂ Melting Pt: 73…75 °C |
MDL Number:
MFCD00794701 CAS Number: 339016-47-6 |
Specification Test Results
Melting point | 73-75°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...