4-Chloro-N-(1,3-thiazol-2-yl)butanamide ≥95%
Supplier: Matrix Scientific
045308-500MG
045308-1G
045308-5G
101870-228EA
232.31
USD
101870-228
101870-226
101870-230
4-Chloro-N-(1,3-thiazol-2-yl)butanamide ≥95%
4-Chloro-N-(1,3-thiazol-2-yl)butanamide
Hazard: Irritant
Formula:
C₇H₉ClN₂OS Melting Pt: 142…145 °C |
MDL Number:
MFCD00664607 CAS Number: 37762-99-5 |
Specification Test Results
Melting point | 142-145°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...