5-Methyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazol-4-ol ≥95%

Supplier: Matrix Scientific

045252-500MG
101869-936EA 556.92 USD
101869-936
5-Methyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazol-4-ol ≥95%
5-Methyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazol-4-ol

Hazard: Irritant

Formula: C₁₁H₉F₃N₂O
Melting Pt: 156…158 °C
MDL Number: MFCD03426594
CAS Number: 119868-25-6

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR