Methyl-6-methyl-3-[(4-methylbenzoyl)amino]-2-oxo-2H-pyran-5-carboxylate ≥95%

Supplier: Matrix Scientific

045240-500MG 045240-1G 045240-5G
101869-870EA 375.38 USD
101869-870 101869-872 101869-874
Methyl-6-methyl-3-[(4-methylbenzoyl)amino]-2-oxo-2H-pyran-5-carboxylate ≥95%
Methyl-6-methyl-3-[(4-methylbenzoyl)amino]-2-oxo-2H-pyran-5-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₅NO₅
Melting Pt: 181…183 °C
MDL Number: MFCD00139849
CAS Number: 339009-31-3

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR