Ethyl-3-[(3-chlorobenzoyl)amino]-2-oxo-6-phenyl-2H-pyran-5-carboxylate ≥95%
Supplier: Matrix Scientific
045239-1G
045239-500MG
045239-5G
101869-864EA
375.38
USD
101869-864
101869-866
101869-868
Ethyl-3-[(3-chlorobenzoyl)amino]-2-oxo-6-phenyl-2H-pyran-5-carboxylate ≥95%
Ethyl-3-[(3-chlorobenzoyl)amino]-2-oxo-6-phenyl-2H-pyran-5-carboxylate
Hazard: Irritant
Formula:
C₂₁H₁₆ClNO₅ Melting Pt: 181…182 °C |
MDL Number:
MFCD00139848 CAS Number: 339009-30-2 |
Specification Test Results
Melting point | 181-182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...