4-(4-Chlorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile ≥95%
Supplier: Matrix Scientific
045230-500MG
045230-5G
045230-1G
101869-810EA
181.54
USD
101869-810
101869-812
101869-814
4-(4-Chlorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile ≥95%
4-(4-Chlorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile
Hazard: Irritant
Formula:
C₁₁H₆ClNO₂ MW: 219.63 g/mol Melting Pt: 230…232 °C |
MDL Number:
MFCD00099688 CAS Number: 39561-83-6 |
Specification Test Results
Melting point | 230-232°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...