Methyl-4-amino-2-(4-benzylpiperazino)-1,3-thiazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
045227-1G
045227-500MG
101869-796EA
556.92
USD
101869-796
101869-798
Methyl-4-amino-2-(4-benzylpiperazino)-1,3-thiazole-5-carboxylate ≥95%
Methyl-4-amino-2-(4-benzylpiperazino)-1,3-thiazole-5-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₀N₄O₂S MW: 332.43 g/mol Melting Pt: 172…174 °C |
MDL Number:
MFCD00139424 CAS Number: 343375-51-9 |
Specification Test Results
Melting point | 172-174°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...