Ethyl-3-(4-methoxyphenyl)acrylate ≥97%
Supplier: Matrix Scientific
045200-1G
045200-5G
045200-500MG
101869-660EA
232.31
USD
101869-660
101869-662
101869-658
Ethyl-3-(4-methoxyphenyl)acrylate ≥97%
Ethyl-3-(4-methoxyphenyl)acrylate
Hazard: Irritant
Formula:
C₁₂H₁₄O₃ Boiling Pt: 177…180 °C (15 Torr) Melting Pt: 50…52 °C |
MDL Number:
MFCD00026906 CAS Number: 1929-30-2 |
Specification Test Results
Melting point | 50-52°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...