3-(1,3-Benzodioxol-5-yl)-3-(1,4-dioxa-8-azaspiro[4.5]dec-8-yl)propanoic acid ≥95%

Supplier: Matrix Scientific

045199-500MG 045199-1G 045199-5G
101869-652EA 278.46 USD
101869-652 101869-654 101869-656
3-(1,3-Benzodioxol-5-yl)-3-(1,4-dioxa-8-azaspiro[4.5]dec-8-yl)propanoic acid ≥95%
3-(1,3-Benzodioxol-5-yl)-3-(1,4-dioxa-8-azaspiro[4.5]dec-8-yl)propanoic acid

Hazard: Irritant

Formula: C₁₇H₂₁NO₆
Melting Pt: 191…193 °C
MDL Number: MFCD08056651
CAS Number: 886361-92-8

Order Now

Specification Test Results

Melting point 191-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR